Learn More
Chemglass Life Sciences Distilling Column, 24/40 Joint, 25 x 300mm, Hemple
SDP
Catalog No. CG122811
Encompass_Preferred
Distilling Column, 24/40 Joint, 25 x 300mm, Hemple. Plain distillation column with a row of indentations to support column packing material. Column length listed is for the straight portion above the row of indentations and below the joint. Column has a standard taper top outer and lower inner joint.
Catalog No. CG122811 Supplier Chemglass Life Sciences Supplier No. CG122811
Distilling Column, 24/40 Joint, 25 x 300mm, Hemple. Plain distillation column with a row of indentations to support column packing material. Column length listed is for the straight portion above the row of indentations and below the joint. Column has a standard taper top outer and lower inner joint.