Learn More
Chemglass Life Sciences Saddles, Berl, 1000cc, Porcelain, Distilling Column, Approx. 1250 Pieces/Package
SDP
Catalog No. CG128402
Encompass_Preferred
Saddles, Berl, 1000cc, Porcelain, Distilling Column, Approx. 1250 Pieces/Package. Porcelain berl saddles having an O.D. of 8mm. Ideal for vacuum distillation, absorption and extraction. Saddles provide a highly useable surface area, lower gas flow resistance and excellent internal distribution. Available in packages of 200cc and 1000cc boxes.
Catalog No. CG128402 Supplier Chemglass Life Sciences Supplier No. CG128402
Saddles, Berl, 1000cc, Porcelain, Distilling Column, Approx. 1250 Pieces/Package. Porcelain berl saddles having an O.D. of 8mm. Ideal for vacuum distillation, absorption and extraction. Saddles provide a highly useable surface area, lower gas flow resistance and excellent internal distribution. Available in packages of 200cc and 1000cc boxes.