Learn More
Chemglass Life Sciences Distilling Column, 14/20 Joint, 100mm, Vigreux
SDP
Supplier:  Chemglass Life Sciences CG123110
Encompass_Preferred
Distilling Column, 14/20 Joint, 100mm, Vigreux. Vigreux style column with indentations that run the entire length of the column for improved vapor-liquid contact. Column length refers to the effective length, measured from the lowest to highest indent in the column. Column has a top outer and lower inner standard taper joint.
Catalog No. CG123110