Learn More
Chemglass Life Sciences Distilling Column, 24/40 Joint, 150mm, Vigreux
SDP
Catalog No. CG123116
Encompass_Preferred
Distilling Column, 24/40 Joint, 150mm, Vigreux. Vigreux style column with indentations that run the entire length of the column for improved vapor-liquid contact. Column length refers to the effective length, measured from the lowest to highest indent in the column. Column has a top outer and lower inner standard taper joint.
Catalog No. CG123116 Supplier Chemglass Life Sciences Supplier No. CG123116
Distilling Column, 24/40 Joint, 150mm, Vigreux. Vigreux style column with indentations that run the entire length of the column for improved vapor-liquid contact. Column length refers to the effective length, measured from the lowest to highest indent in the column. Column has a top outer and lower inner standard taper joint.