Learn More
Wilmad Labglass DIST HEAD 200MM 24/40
Catalog No. 50805426
Encompass_Preferred
Distilling Head. Jacket Length: 200mm; Joint Size: [ST] 24/40; Thermometer Joint: [ST] 10/30. One piece design w/ Condenser angled at 75degree Designed w/ a vacuum port and For jointed thermometers w/ a 75mm immersion length.
Catalog No. 50-805-426 Supplier Wilmad Labglass Supplier No. LG6115100
Distilling Head. Jacket Length: 200mm; Joint Size: [ST] 24/40; Thermometer Joint: [ST] 10/30. One piece design w/ Condenser angled at 75degree Designed w/ a vacuum port and For jointed thermometers w/ a 75mm immersion length.