Learn More
Wilmad Labglass DIST HEAD 400MM 29/42
Supplier:  Wilmad Labglass LG6115126
Encompass_Preferred
Distilling Head. Jacket Length: 400mm; Joint Size: [ST] 29/42; Thermometer Joint: [ST] 10/30. One piece design w/ Condenser angled at 75degree Designed w/ a vacuum port and For jointed thermometers w/ a 75mm immersion length.
Catalog No. 50-805-160