Learn More
Wilmad Labglass DIST.HEAD MCRO ST19/22
Catalog No. 50805040
Encompass_Preferred
Distilling Head. Joint Size: [ST] 19/22; Jacket Length: 100mm. One piece design w/ the Liebig Condenser angled at 75degree Receiver joint is angled downward and has hose connection For connection to vacuum source.
Catalog No. 50-805-040 Supplier Wilmad Labglass Supplier No. ML870702
Distilling Head. Joint Size: [ST] 19/22; Jacket Length: 100mm. One piece design w/ the Liebig Condenser angled at 75degree Receiver joint is angled downward and has hose connection For connection to vacuum source.