Learn More
Chemglass Life Sciences Distilling Head, Short Path, Vigreux Indents, 10/30 Thermometer Joint, 24/40 Lower Inner Joint, Approx. 195mm Width x 160mm Height
SDP
Catalog No. CG123912
Encompass_Preferred
Distilling Head, Short Path, Vigreux Indents, 10/30 Thermometer Joint, 24/40 Lower Inner Joint, Approx. 195mm Width x 160mm Height. Designed for use with small quantities where hold up between the flask and receiver must be kept to a minimum. The thermometer bulb and stem serve as column packing. Upper hose connections are for cooling lines while the lower connection is for vacuum. Available with Vigreux indentations to improve efficiency. NOTE: The 14/20 size accepts a 1” immersion 7/12 or 10/18 thermometer, while the 24/40 and 29/42 sizes accepts a 2” immersion, 10/30 thermometer.
Catalog No. CG123912 Supplier Chemglass Life Sciences Supplier No. CG123912
Distilling Head, Short Path, Vigreux Indents, 10/30 Thermometer Joint, 24/40 Lower Inner Joint, Approx. 195mm Width x 160mm Height. Designed for use with small quantities where hold up between the flask and receiver must be kept to a minimum. The thermometer bulb and stem serve as column packing. Upper hose connections are for cooling lines while the lower connection is for vacuum. Available with Vigreux indentations to improve efficiency. NOTE: The 14/20 size accepts a 1” immersion 7/12 or 10/18 thermometer, while the 24/40 and 29/42 sizes accepts a 2” immersion, 10/30 thermometer.