Learn More
Chemglass Life Sciences Distilling Head, Short Path, Jacketed, 10/18 Thermometer Joint, 14/20 Lower Inner Joint, Approx. 105mm Width x 105mm Height
SDP
Catalog No. CG124005
Encompass_Preferred
Distilling Head, Short Path, Jacketed, 10/18 Thermometer Joint, 14/20 Lower Inner Joint, Approx. 105mm Width x 105mm Height. Similar to CG-1239 but vacuum jacketed to improve efficiency. Available with Vigreux indentations in uptake tube to further increase the performance of the head. NOTE: The 14/20 size accepts a 1” immersion 7/12 or 10/18 thermometer, while the 24/40 and 29/42 sizes accept a 2” immersion, 10/30 thermometer.
Catalog No. CG124005 Supplier Chemglass Life Sciences Supplier No. CG124005
Distilling Head, Short Path, Jacketed, 10/18 Thermometer Joint, 14/20 Lower Inner Joint, Approx. 105mm Width x 105mm Height. Similar to CG-1239 but vacuum jacketed to improve efficiency. Available with Vigreux indentations in uptake tube to further increase the performance of the head. NOTE: The 14/20 size accepts a 1” immersion 7/12 or 10/18 thermometer, while the 24/40 and 29/42 sizes accept a 2” immersion, 10/30 thermometer.