Learn More
Chemglass Life Sciences Distilling Head, Short Path, Improved, Vigreux Indents, 10/30 Thermometer Joint, 24/40 Lower Inner Joint, Approx. 195mm Width x 160mm Height
SDP
Catalog No. CG1239A03
Encompass_Preferred
Distilling Head, Short Path, Improved, Vigreux Indents, 10/30 Thermometer Joint, 24/40 Lower Inner Joint, Approx. 195mm Width x 160mm Height. Similar to CG-1239, but with the condenser being a combination of a cold coil and Liebig. Water flows into hose connection “A” through the coil and exits the outer jacket through hose connection “B”. Head is excellent for use when distilling medium to high boiling point solvents. Top outer joint is a 10/30 for use with a 2” immersion thermometer. Available with or without Vigreux indentations to further improve efficiency.
Catalog No. CG1239A03 Supplier Chemglass Life Sciences Supplier No. CG1239A03
Distilling Head, Short Path, Improved, Vigreux Indents, 10/30 Thermometer Joint, 24/40 Lower Inner Joint, Approx. 195mm Width x 160mm Height. Similar to CG-1239, but with the condenser being a combination of a cold coil and Liebig. Water flows into hose connection “A” through the coil and exits the outer jacket through hose connection “B”. Head is excellent for use when distilling medium to high boiling point solvents. Top outer joint is a 10/30 for use with a 2” immersion thermometer. Available with or without Vigreux indentations to further improve efficiency.