Learn More
Wilmad Labglass DIST HEAD SOLV STILL 500ML
Supplier:  Wilmad Labglass LG6330T100
Encompass_Preferred
Distilling Head, Solvent Purification. Storage Capacity: 500mL; Flask Size: 1000mL; Joint Size: [ST] 24/40; Side Take-off Arm Joint; [ST] 14/20. Flask provides storage and access by syringe
Catalog No. 50-805-541