Learn More
Chemglass Life Sciences Distillation Head, Vigruex, Jacketed, 10/18 Thermometer Joint, 14/20 Joint Size, 100mm Column Length, Approx. 200mm Height x 180mm Width
SDP
Catalog No. CG124610
Encompass_Preferred
Distillation Head, Vigruex, Jacketed, 10/18 Thermometer Joint, 14/20 Joint Size, 100mm Column Length, Approx. 200mm Height x 180mm Width. Vacuum jacketed Vigreux distillation head having a sealed on condenser, braced for strength. The top outer joint is for a 1” immersion 10/18 or 10/30 thermometer. Vacuum adapter has an inner joint and a sealed in drip tip. Condenser length is 100mm on the 14/20 size and 200mm on the 24/40 and 29/42 sizes.
Catalog No. CG124610 Supplier Chemglass Life Sciences Supplier No. CG124610
Distillation Head, Vigruex, Jacketed, 10/18 Thermometer Joint, 14/20 Joint Size, 100mm Column Length, Approx. 200mm Height x 180mm Width. Vacuum jacketed Vigreux distillation head having a sealed on condenser, braced for strength. The top outer joint is for a 1” immersion 10/18 or 10/30 thermometer. Vacuum adapter has an inner joint and a sealed in drip tip. Condenser length is 100mm on the 14/20 size and 200mm on the 24/40 and 29/42 sizes.