Learn More
Chemglass Life Sciences Distilling Receiver, Complete, 15mL, 14/20, Approx. 220mm Height x 90mm Width
SDP
Catalog No. CG128114
Encompass_Preferred
Distilling Receiver, Complete, 15mL, 14/20, Approx. 220mm Height x 90mm Width. Similar to CG-1279 but with the top outer joint vertical instead of a 105º angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.
Catalog No. CG128114 Supplier Chemglass Life Sciences Supplier No. CG128114
Distilling Receiver, Complete, 15mL, 14/20, Approx. 220mm Height x 90mm Width. Similar to CG-1279 but with the top outer joint vertical instead of a 105º angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.