Learn More
Chemglass Life Sciences Distilling Receiver, Complete, 50mL, 24/40, Approx. 449mm Height x 160mm Width
SDP
Catalog No. CG128101
Encompass_Preferred
Distilling Receiver, Complete, 50mL, 24/40, Approx. 449mm Height x 160mm Width. Similar to CG-1279 but with the top outer joint vertical instead of a 105º angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.
Catalog No. CG128101 Supplier Chemglass Life Sciences Supplier No. CG128101
Distilling Receiver, Complete, 50mL, 24/40, Approx. 449mm Height x 160mm Width. Similar to CG-1279 but with the top outer joint vertical instead of a 105º angle. Complete item consists of a vacuum adapter, distribution adapter, four of the listed size flasks with hooks and ten CG-110 stainless steel springs.