Learn More
Chemglass Life Sciences Distilling Receiver, 20mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 325mm Height x 95mm Width
SDP
Supplier:  Chemglass Life Sciences CG126127
Encompass_Preferred
Distilling Receiver, 20mL, Moisture Test, Barrett, 24/40 Joint Size, 2mm PTFE Stopcock, Approx. 325mm Height x 95mm Width. Barrett style distilling receiver used to determine the water content in petroleum or bituminous material. Receiver is made according to ASTM specifications, with the addition of the top outer standard taper joint and lower stopcock. The 2mL has sub-divisions of 0.02mL; the 5ml has subdivisions of 0.20mL; 10ml has subdivisions of 0.1mL; the 20mL has subdivisions of 0.2 from 0 to 3mL and 0.5 from 3 to 20mL. Available with glass or PTFE stopcocks.
Catalog No. CG126127