Learn More
Chemglass Life Sciences Distilling Receiver, 10mL, Moisture Test, Barrett, 14/20 Joint Size, 2mm PTFE Stopcock , Approx. 190mm Height x 75mm Width
SDP
Catalog No. CG126221
Encompass_Preferred
Distilling Receiver, 10mL, Moisture Test, Barrett, 14/20 Joint Size, 2mm PTFE Stopcock , Approx. 190mm Height x 75mm Width. Similar to CG-1261 but with the uptake tube vacuum jacketed to improve distillation performance. The jacket eliminates the possibility of condensation or recrystallization of distillate in the sidearm. The 5ml has subdivisions of 0.20ml; 10ml has subdivisions of 0.1ml; the 20ml has subdivisions of 0.2 from 0 to 3ml and 0.5 from 3 to 20mL. Available with glass or PTFE stopcocks.
Catalog No. CG126221 Supplier Chemglass Life Sciences Supplier No. CG126221
Distilling Receiver, 10mL, Moisture Test, Barrett, 14/20 Joint Size, 2mm PTFE Stopcock , Approx. 190mm Height x 75mm Width. Similar to CG-1261 but with the uptake tube vacuum jacketed to improve distillation performance. The jacket eliminates the possibility of condensation or recrystallization of distillate in the sidearm. The 5ml has subdivisions of 0.20ml; 10ml has subdivisions of 0.1ml; the 20ml has subdivisions of 0.2 from 0 to 3ml and 0.5 from 3 to 20mL. Available with glass or PTFE stopcocks.