Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: ABO/Rh Blood Typing
GREENER_CHOICESCI_ED

Catalog No. S100903 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S100903 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S100903 Supplier Innovating Science™ Supplier No. IS3110
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This educational kit provides the most procedurally accurate simulation of the ABO/Rh blood typing technique available.

  • In this kit, students test and determine the ABO/Rh blood types of four different simulated blood samples using Innovating Science's new simulated blood.
  • Students simply combine the blood samples and antisera, gently agitate the blood typing tray, and observe the results – no toothpicks, no stirring, and no waiting for results required.
  • Kit contains an Instruction Manual and enough materials for 2 groups of students.

Specifications

Grade 11 to 14
Includes Simulated blood and simulated antisera
Material Plastic Bottles
Class Size 2 Lab Groups
Product Type Forensic Kit
Description Disposable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.