Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Elephant Toothpaste
GREENER_CHOICESCI_ED

Catalog No. S08192 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S08192 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08192 Supplier Innovating Science™ Supplier No. IS1501
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Teaches students the effects of a catalyst on a chemical reaction

  • Teach students the effect of a catalyst on a chemical reaction.
  • Displays decomposition of hydrogen peroxide by mixing dish soap and adding yeast (catalyst) to create foam.
  • Includes:
    • 50 mL hydrogen peroxide (3%)
    • 6 mL dish soap
    • 2 g yeast
    • 2 plastic cups, 5 oz.
    • 2 stir sticks
    • 1 clear bottle, 8 oz.
  • Kit contains an instruction manual and enough materials to perform the demonstration twice.

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.