Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Paper Chromatography
GREENER_CHOICESCI_ED

Catalog No. S08193 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S08193 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08193 Supplier Innovating Science™ Supplier No. IS1502
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Students perform paper chromatography on three dyes, a dye mixture, and determine the mixture's composition

  • Objectives:
    • Perform paper chromatography on three dyes
    • Perform paper chromatography on a dye mixture
    • Determine mixture composition
    • Understand component of a chromatography system and why different substances separate/move at different rates
  • One dye set includes:
    • 0.5 mL crystal violet
    • 0.5 mL safranin O
    • 0.5 mL toluidine blue
    • 0.5 mL chromatography mixture
    • 30 mL 50% ethanol
    • 2 chromatography sheets
    • 4 capillary tubes
    • 2 plastic cups, 10 oz.
  • Notes:
    • Includes an instruction manual and enough materials for two groups

Specifications

Grade 7 to 15
Age 12 to 20
Class Size 2 Lab Groups
Product Type Chromatography Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.