Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Distance Learning: Periodic Table- Nonmetals, Metals, and Metalloids
GREENER_CHOICESCI_ED

Catalog No. S08195 Shop All Aldon Products
Change view
Click to view available options
Class Size:
2 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S08195 2 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S08195 Supplier Innovating Science™ Supplier No. IS1504
Add to Cart
Add to Cart

Students examine the physical properties of nonmetals, metals, and metalloids as well as chemical reactivity

  • Objectives:
    • Learn about Dmitri Mendeleev's construction of the periodic table
    • Periodic table organization is based on properties of elements in relation to others
    • Examine physical properties of several elements
    • Test chemical reactivity of elements
    • Use knowledge to classify each tested element as nonmetal, metal, or metalloid
  • Includes:
    • 4 g aluminum pellets
    • 4 g carbon pellets
    • 4 g silicon pellets
    • 4 g zinc pieces
    • 4 g sulfur pieces
    • 5 g sodium bicarbonate
    • 6 copper pieces
    • 6 magnesium pieces
    • 20 mL hydrochloric acid 1 N
    • 20 mL copper (II) chloride
    • 14 test tubes
    • 4 pipettes
    • 2 nails
    • 6 plastic cups, 5 oz.
    • 1 plastic scoop
  • Notes:
    • Kit contains instruction manual and enough materials for two groups of students

Specifications

Grade 7 to 15
Age 12 to 20
Content And Storage Ambient
Class Size 2 Lab Groups
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.