Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medicom™ DisTech™ Earloop Face Mask, Pleated, Nonsterile

Catalog No. 1288830 Shop All Medicom Products
Change view
Click to view available options
Earloops
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
12-888-30 Earloops
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-888-30 Supplier Medicom™ Supplier No. 2215

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Primary protection for simple, low-risk procedures involving very light dust, moisture, spray, and spatter. Adjustable nosepiece for custom fit.

  • Inner and outer layer constructed for basic fluid resistance
  • Ultrasonically sealed, soft, stretchy, latex free earloops
  • Adjustable nosepiece for custom fit and comfortable, easy breathing
  • Designed for use during medical procedures to protect both the patient and the wearer from transfer of microorganisms, body fluids and particulate matter
  • Economic alternative for simple procedures
  • Packaging: 50 masks/box; 40 boxes/case for a total of 2000 masks/case
  • User: Adult

Specifications

Product Type Medical/Procedural Face Mask
ASTM Level ASTM Level 1
Color Blue
Fastening Type Earloops
Material Spunbond Polypropylene
Size Standard
Sterility Non-sterile
Style Pleated
Disposable Single-use
Filter Efficiency ≥95% (PFE), ≥95% (BFE)
For Use With (Application) Low-risk procedures involving very light dust, moisture, spray and splatter
Latex Free Yes
Model 2215
Packaging Quantity 2000/Cs. (50/box; 40 boxes/cs.)
Resistance Fluid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.