Learn More
MyBioSource Rabbit distemper virus Fusion glycoprotein F0 Polyclonal Antibody, 0.1 mg
SDP
Supplier:  MyBioSource MBS14984580.1 MG
Encompass_Preferred
Class I viral fusion protein. 95%, Protein G purified. Polyclonal, IgG, Rabbit.
Catalog No. NC1566845