Learn More
Chemglass Life Sciences Distilling Column, 14/20 Joint, 10 x 130mm, Hemple
SDP
Catalog No. CG122805
Encompass_Preferred
Distilling Adapter, Double, 10° Angled Side Neck, 24/40 Joint, 6mm PTFE Stopcock, 1/2" Swagelok. Adapter is designed for use with flasks or reaction vessel lids that have a 10° angled side neck. Adapter has a lower inner joint and two top outer joints. Lower stopcock is a 6mm PTFE stopcock with a 1/2” or 3/4” OD distillate takeoff for use with a compression fitting and TFE tubing. PLEASE NOTE: ADAPTER CAN BE USED WITH COMPOUND ANGLED SIDE NECKS.
Catalog No. CG122805 Supplier Chemglass Life Sciences Supplier No. CG122805
Distilling Adapter, Double, 10° Angled Side Neck, 24/40 Joint, 6mm PTFE Stopcock, 1/2" Swagelok. Adapter is designed for use with flasks or reaction vessel lids that have a 10° angled side neck. Adapter has a lower inner joint and two top outer joints. Lower stopcock is a 6mm PTFE stopcock with a 1/2” or 3/4” OD distillate takeoff for use with a compression fitting and TFE tubing. PLEASE NOTE: ADAPTER CAN BE USED WITH COMPOUND ANGLED SIDE NECKS.