Learn More
Chemglass Life Sciences Solvent Distillation Apparatus Complete, Airfree, Schlenk
SDP
Catalog No. AF074001
Encompass_Preferred
Solvent Distillation Apparatus Complete, Airfree, Schlenk. Similar in design to AF0720 but without delivery adapter. Lower 3-way valve has been replaced with a single 0-4mm Chem-Vac™ Chem-Cap valve for draining distillate back into the flask. The uptake tube has one row of vigreux indentations, while the 14/20 top outer joint has been eliminated. Temperature can be measured by using a 2-neck flask with a thermocouple or thermometer adapter. The delivery tube has two 2mm PTFE straight bore stopcocks that can be operated in a way to purge the receiving vessel with inert gas. Capacity of solvent storage area is approx 250mL with a 24/40 top outer and a 24/40 lower inner standard taper joints.Design suggested by Dr. Zoltan Tyeklar, Metasyn, Inc, Cambridge, MA
Catalog No. AF074001 Supplier Chemglass Life Sciences Supplier No. AF074001
Solvent Distillation Apparatus Complete, Airfree, Schlenk. Similar in design to AF0720 but without delivery adapter. Lower 3-way valve has been replaced with a single 0-4mm Chem-Vac™ Chem-Cap valve for draining distillate back into the flask. The uptake tube has one row of vigreux indentations, while the 14/20 top outer joint has been eliminated. Temperature can be measured by using a 2-neck flask with a thermocouple or thermometer adapter. The delivery tube has two 2mm PTFE straight bore stopcocks that can be operated in a way to purge the receiving vessel with inert gas. Capacity of solvent storage area is approx 250mL with a 24/40 top outer and a 24/40 lower inner standard taper joints.Design suggested by Dr. Zoltan Tyeklar, Metasyn, Inc, Cambridge, MA