Learn More
Chemglass Life Sciences 500mL Solvent Distillation Apparatus Complete, Airfree, Schlenk
SDP
Catalog No. AF072002
Encompass_Preferred
500mL Solvent Distillation Apparatus Complete, Airfree, Schlenk. Apparatus for use in distillation of materials like THF or ether. Storage reservoir has a capacity of 500mL. The cold finger coil condenser has a section that's immersed into the distillate. The 0-4mm Chem-Vac™ Chem-Cap valves provide for grease-free, leakproof seals. Side vapor tube is fitted w/a 14/20 standard taper outer joint, to be used w/a CG104214 thermometer adapter which fits a plain stem thermometer. A short (50mm) vigreux section is provided to enhance separation. Syringe access port incorporates a 4mm PTFE stopcock w/a 14/20 outer joint for use w/CG-3022 septum stopper. Lower 3way valve allows for solvent return to flask, takeoff or storage in graduated reservoir. Delivery adapter is fitted w/a 12/5 spherical joint for flexibility & a 0-4mm Chem-Vac™ Chem-Cap valve for purging of the receiving vessel.
Catalog No. AF072002 Supplier Chemglass Life Sciences Supplier No. AF072002
500mL Solvent Distillation Apparatus Complete, Airfree, Schlenk. Apparatus for use in distillation of materials like THF or ether. Storage reservoir has a capacity of 500mL. The cold finger coil condenser has a section that's immersed into the distillate. The 0-4mm Chem-Vac™ Chem-Cap valves provide for grease-free, leakproof seals. Side vapor tube is fitted w/a 14/20 standard taper outer joint, to be used w/a CG104214 thermometer adapter which fits a plain stem thermometer. A short (50mm) vigreux section is provided to enhance separation. Syringe access port incorporates a 4mm PTFE stopcock w/a 14/20 outer joint for use w/CG-3022 septum stopper. Lower 3way valve allows for solvent return to flask, takeoff or storage in graduated reservoir. Delivery adapter is fitted w/a 12/5 spherical joint for flexibility & a 0-4mm Chem-Vac™ Chem-Cap valve for purging of the receiving vessel.