Learn More
Chemglass Life Sciences Distilling Column, Water Jacketed, 19/22 Joint, 200mm
SDP
Catalog No. CG123008
Encompass_Preferred
Distilling Column, Water Jacketed, 19/22 Joint, 200mm. Jacketed distilling column has indentations on the inner tube to support packing material. The jacket allows the column to be used as a condenser. Column has a top outer and lower inner standard taper joint. Available in joint sizes other than those listed. All sizes have an I.D. of approximately 10mm.
Catalog No. CG123008 Supplier Chemglass Life Sciences Supplier No. CG123008
Distilling Column, Water Jacketed, 19/22 Joint, 200mm. Jacketed distilling column has indentations on the inner tube to support packing material. The jacket allows the column to be used as a condenser. Column has a top outer and lower inner standard taper joint. Available in joint sizes other than those listed. All sizes have an I.D. of approximately 10mm.