Learn More
Chemglass Life Sciences Chemglass Side Support Distillation Shelf with PTFE Ring for 30-50 L Reactor Stands
SDP
Catalog No. NC2824283
Encompass_Preferred
Removable side support shelf for 30-50 L reactor stands, designed to support distillation receiving flasks. The shelf features a PTFE flask support ring and PTFE-coated aluminum shelf for chemical resistance, a V-shaped cutout for easy flask insertion and viewing, and mounts to the uprights of reactor support frames so it can be positioned on either side.
  • PTFE flask support ring for chemical resistance.
  • PTFE-coated aluminum shelf for corrosion resistance.
  • Supports 10-20 L round-bottom flasks.
  • Dimensions: 12-7/8 in W x 13 in L.
  • Mounts to uprights and can be installed on either side of the frame.
  • V-shaped cutout for easy flask insertion and viewing.
  • Provides stable support for distillation receiving flasks during processing.
Catalog No. NC2824283 Supplier Chemglass Life Sciences Supplier No. CG1963D50
Removable side support shelf for 30-50 L reactor stands, designed to support distillation receiving flasks. The shelf features a PTFE flask support ring and PTFE-coated aluminum shelf for chemical resistance, a V-shaped cutout for easy flask insertion and viewing, and mounts to the uprights of reactor support frames so it can be positioned on either side.
  • PTFE flask support ring for chemical resistance.
  • PTFE-coated aluminum shelf for corrosion resistance.
  • Supports 10-20 L round-bottom flasks.
  • Dimensions: 12-7/8 in W x 13 in L.
  • Mounts to uprights and can be installed on either side of the frame.
  • V-shaped cutout for easy flask insertion and viewing.
  • Provides stable support for distillation receiving flasks during processing.