Learn More
Chemglass Life Sciences Support Distillation Cart, Dual, 10L to 20L
SDP
Catalog No. 501531318
Encompass_Preferred
Support Distillation Cart, Dual, 10L to 20L. Distillation receiver carts for use with Chemglass's 10L thru 100L reactor systems. Heavy Duty support cart can be used for round bottom flasks or cylindrical distillation receivers. Rear of cart incorporates 1/2 Stainless Steel lattice for mounting support clamps or accessories. Glassware can be quickly installed and removed by one person.
Catalog No. 50-153-1318 Supplier Chemglass Life Sciences Supplier No. CG1968D102
Support Distillation Cart, Dual, 10L to 20L. Distillation receiver carts for use with Chemglass's 10L thru 100L reactor systems. Heavy Duty support cart can be used for round bottom flasks or cylindrical distillation receivers. Rear of cart incorporates 1/2 Stainless Steel lattice for mounting support clamps or accessories. Glassware can be quickly installed and removed by one person.