Learn More
Chemglass Life Sciences Distilling Column, Snyder, 24/40 Joint, 150mm, Improved
SDP
Catalog No. CG130401
Encompass_Preferred
Distilling Column, Snyder, 24/40 Joint, 150mm, Improved. Three section Snyder column with improved design to prevent column flooding. Column has a 24/40 top outer joint and a 24/40 lower inner joint for use with CG-1308 Kuderna-Danish flask.
Catalog No. CG130401 Supplier Chemglass Life Sciences Supplier No. CG130401
Distilling Column, Snyder, 24/40 Joint, 150mm, Improved. Three section Snyder column with improved design to prevent column flooding. Column has a 24/40 top outer joint and a 24/40 lower inner joint for use with CG-1308 Kuderna-Danish flask.