Learn More
Chemglass Life Sciences Distilling Receiver, 250mL, Solvent Recovery, Graduated, 24/40 Joint Size, 2mm PTFE Stopcock, 4mm 3-Way PTFE Stopcock, Approx. 415mm Height x 185mm Width
SDP
Catalog No. CG127201
Encompass_Preferred
Distilling Receiver, 250mL, Solvent Recovery, Graduated, 24/40 Joint Size, 2mm PTFE Stopcock, 4mm 3-Way PTFE Stopcock, Approx. 415mm Height x 185mm Width. Graduated distillation receiver with a 4mm three way stopcock to permit solvent removal through the 10mm O.D. tube or return to distillation flask. Upper sampling port has a 2mm PTFE stopcock. The top outer and lower inner joints are the same size.
Catalog No. CG127201 Supplier Chemglass Life Sciences Supplier No. CG127201
Distilling Receiver, 250mL, Solvent Recovery, Graduated, 24/40 Joint Size, 2mm PTFE Stopcock, 4mm 3-Way PTFE Stopcock, Approx. 415mm Height x 185mm Width. Graduated distillation receiver with a 4mm three way stopcock to permit solvent removal through the 10mm O.D. tube or return to distillation flask. Upper sampling port has a 2mm PTFE stopcock. The top outer and lower inner joints are the same size.