Learn More
Ace Glass, Inc. Distillation head, auto reflux control, complete
SDP
Catalog No. NC0606350
Encompass_Preferred
A small, low-holdup automatic reflux control distilling head which can be completely disassembled for easy cleaning or replacement of parts. Lower column joint and side arm for condenser are 24/40. Takeoff arm joint is 28/11, plunger guide is 19/38, and thermometer joint is 10/30.
Catalog No. NC0606350 Supplier Ace Glass, Inc. Supplier No. 659810
A small, low-holdup automatic reflux control distilling head which can be completely disassembled for easy cleaning or replacement of parts. Lower column joint and side arm for condenser are 24/40. Takeoff arm joint is 28/11, plunger guide is 19/38, and thermometer joint is 10/30.