Learn More
Synthware™ Distillation Head- Microscale
GREENER_CHOICESDP
Catalog No. 31501061 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
10/18, 19/22
Catalog No. Standard Taper Joint
31-501-061 10/18, 19/22
1 options
Catalog No. 31-501-061 Supplier Synthware™ Supplier No. D112440G
Add to Cart
Add to Cart
With 2 mm glass stopcock and 24/40 joint. Supplied with a special 25 mL long-neck flask for small volume efficiency.
Specifications
For Use With (Application) Organic Synthesis
Type Vacuum Distillation Apparatus
Material Borosilicate Glass
For Use With (Equipment) Thermometer
Quantity 1 Ea.
Standard Taper Joint 10/18, 19/22
Joint Type Ground Glass