Learn More
Chemglass Life Sciences Distillation Head, Micro, Vigruex, 10/18 Thermometer Joint, 14/20 Outer Joint, Jacketed, Approx. 195mm Height x 185mm Width
SDP
Catalog No. CG124210
Encompass_Preferred
Distillation Head, Micro, Vigruex, 10/18 Thermometer Joint, 14/20 Outer Joint, Jacketed, Approx. 195mm Height x 185mm Width. Distillation head with condenser sealed to a Vigreux column. Top thermometer joint is a 10/18 for use with a 1¿ immersion thermometer. Vacuum adapter has an outer joint and accepts a CG-1279-11 distribution adapter. Similar to CG-1241 but with the Vigreux column vacuum jacketed to improve distillation performance.
Catalog No. CG124210 Supplier Chemglass Life Sciences Supplier No. CG124210
Distillation Head, Micro, Vigruex, 10/18 Thermometer Joint, 14/20 Outer Joint, Jacketed, Approx. 195mm Height x 185mm Width. Distillation head with condenser sealed to a Vigreux column. Top thermometer joint is a 10/18 for use with a 1¿ immersion thermometer. Vacuum adapter has an outer joint and accepts a CG-1279-11 distribution adapter. Similar to CG-1241 but with the Vigreux column vacuum jacketed to improve distillation performance.