Learn More
Ace Glass, Inc. DISTILLATION HEAD W COND JCKT DISTILLATION HEAD W COND JCKT
SDP
Catalog No. NC0250210
Encompass_Preferred
14/20 200mm distillation head with condenser jacket on side arm. Thermometer joint is 10/30. Side arm is at a 75 degree angle.
Catalog No. NC0250210 Supplier Ace Glass, Inc. Supplier No. 935906
14/20 200mm distillation head with condenser jacket on side arm. Thermometer joint is 10/30. Side arm is at a 75 degree angle.