Learn More
Chemglass Life Sciences Distillation Kit, 30/50L Side Shelf, with 10L Receiving Flask
SDP
Catalog No. 5000980682
Encompass_Preferred
Distillation kits for Chemglass 30 and 50L reactor systems. Supplied complete with support shelf to fit CG-1968-X style frame and the components listed in the link below. This add-on kit makes it easy to convert any ofstandard reactors over to a complete distillation system. Please Note: Reactor System Must Be Purchased Separately.
Catalog No. 50-009-80682 Supplier Chemglass Life Sciences Supplier No. CG1968D410
Distillation kits for Chemglass 30 and 50L reactor systems. Supplied complete with support shelf to fit CG-1968-X style frame and the components listed in the link below. This add-on kit makes it easy to convert any ofstandard reactors over to a complete distillation system. Please Note: Reactor System Must Be Purchased Separately.