Learn More
Chemglass Life Sciences Chemglass Distillation Kit for 30/50 L Process Reactor Systems, Side Shelf with 20 L Receiving Flask (CG-1968-D-420)
SDP
Catalog No. NC3344456
Encompass_Preferred
The Chemglass Distillation Kit (CG-1968-D-420) converts 30 L and 50 L process reactor systems into a complete distillation setup. Supplied with a side support shelf designed to fit compatible reactor support frames and a 20 L receiving flask. Reactor system sold separately.
  • Converts standard 30 L and 50 L reactors for distillation use.
  • Includes a side support shelf with PTFE ring for secure mounting.
  • Supplied with a 20 L receiving flask for condensate collection.
  • Supports 10-20 L round-bottom receiving flasks.
  • Side shelf dimensions approximately 12-7/8 in W x 13 in L.
  • Designed for retrofit installation; reactor sold separately.
Catalog No. NC3344456 Supplier Chemglass Life Sciences Supplier No. CG1968D420
Add to Cart
Add to Cart
Request a Quote
The Chemglass Distillation Kit (CG-1968-D-420) converts 30 L and 50 L process reactor systems into a complete distillation setup. Supplied with a side support shelf designed to fit compatible reactor support frames and a 20 L receiving flask. Reactor system sold separately.
  • Converts standard 30 L and 50 L reactors for distillation use.
  • Includes a side support shelf with PTFE ring for secure mounting.
  • Supplied with a 20 L receiving flask for condensate collection.
  • Supports 10-20 L round-bottom receiving flasks.
  • Side shelf dimensions approximately 12-7/8 in W x 13 in L.
  • Designed for retrofit installation; reactor sold separately.