Learn More
Synthware™ Distillation Receiver with Hose Conection
GREENER_CHOICESDP
Catalog No. 31501039 Shop All Kemtech Products
Change view
Click to view available options
Standard Taper Joint:
14/20
19/22
24/40
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Standard Taper Joint
31-501-039 24/40
31-501-037 14/20
31-501-038 19/22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 31-501-039 Supplier Synthware™ Supplier No. D192440
Add to Cart
Add to Cart
Cow-type design has three receivers spaced 40° apart with a 105° angle between top outer joint and lower inner joints.
Specifications
For Use With (Application) Organic Synthesis
Type With Hose Connection
Material Borosilicate Glass
Quantity 1 Ea.
Standard Taper Joint 24/40