Learn More
Chemglass Life Sciences Distillation Support Rod, Stainless Steel, for CG-1968-X Style Frame
SDP
Catalog No. 502733555
Encompass_Preferred
Stainless steel support rod designed to secure distillation assemblies on compatible reactor frames used with 30-50 L process reactor distillation kits. It provides rigid, corrosion-resistant support for condensers and receiving glassware during distillation operations.
  • Stainless steel construction for corrosion resistance.
  • Provides rigid support for condenser and receiving glassware during distillation.
  • Compatible with standard vertical reactor distillation frames; verify fit with your system.
  • Sold individually; availability may vary.
Catalog No. 50-273-3555 Supplier Chemglass Life Sciences Supplier No. CG1968D128
Stainless steel support rod designed to secure distillation assemblies on compatible reactor frames used with 30-50 L process reactor distillation kits. It provides rigid, corrosion-resistant support for condensers and receiving glassware during distillation operations.
  • Stainless steel construction for corrosion resistance.
  • Provides rigid support for condenser and receiving glassware during distillation.
  • Compatible with standard vertical reactor distillation frames; verify fit with your system.
  • Sold individually; availability may vary.