Learn More
Chemglass Life Sciences Distillation Vessel, 250mL, Receiving, Cylindrical, 70mm OD, ChemRxnHub, Compatable with CG-1947-A60
SDP
Catalog No. 50143590
Encompass_Preferred
Distillation Vessel, 250mL, Receiving, Cylindrical, 70mm OD, ChemRxnHub, Compatible with CG-1947-A60. Cylindrical shaped distillation receiving vessel for use with ChemRxnHub benchtop systems for collecting distillate. The top of the vessel has two 0-8 Chem-cap valves with 1/2¿ OD Swagelock side arms that offer a convenient way to connect the receiver to CG-1975 PTFE tubing and compression fittings. Bottom of vessel has 0-14 drain valve with a 1/2¿ OD swagelock sidearm. The body of vessel has approximate volume graduations on the front and back. Vessel body diameter is compatible with CG-1947-A clamps which provide an easy way to support the receiver on the frame or hood lattice.
Catalog No. 50-143-590 Supplier Chemglass Life Sciences Supplier No. CG1267250
Distillation Vessel, 250mL, Receiving, Cylindrical, 70mm OD, ChemRxnHub, Compatible with CG-1947-A60. Cylindrical shaped distillation receiving vessel for use with ChemRxnHub benchtop systems for collecting distillate. The top of the vessel has two 0-8 Chem-cap valves with 1/2¿ OD Swagelock side arms that offer a convenient way to connect the receiver to CG-1975 PTFE tubing and compression fittings. Bottom of vessel has 0-14 drain valve with a 1/2¿ OD swagelock sidearm. The body of vessel has approximate volume graduations on the front and back. Vessel body diameter is compatible with CG-1947-A clamps which provide an easy way to support the receiver on the frame or hood lattice.