Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions DISTILLED DIONIZED WATER 10L
SDP

Catalog No. NC0271937
Encompass_Preferred

Distilled Water Astm Type Ii, Reagent Acs (10l)

Catalog No. NC0271937 Supplier Aqua Solutions Supplier No. 976610LHP
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Distilled Water Astm Type Ii, Reagent Acs (10l)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.