Learn More
Research Products International Corp Distilled Water, Laboratory Reagent Grade, 10 Liter Cubitainer
SDP
Catalog No. 50712781
Encompass_Preferred
Laboratory reagent grade. Purified by steam distillation for use as a research laboratory reagent. Not intended or sold for human consumption. 10 Liter size supplied in a Cubitainer with pouring spout.
Catalog No. 50-712-781 Supplier Research Products International Corp Supplier No. W2052510000.0
Laboratory reagent grade. Purified by steam distillation for use as a research laboratory reagent. Not intended or sold for human consumption. 10 Liter size supplied in a Cubitainer with pouring spout.