Learn More
Chemglass Life Sciences Adapter, Distilling, 24/40 - 10/30, 75mm Immersion, 75°
SDP
Catalog No. CG107601
Encompass_Preferred
Adapter, Distilling, 24/40 - 10/30, 75mm Immersion, 75¿. Adapter is used to connect distilling column with vertical condensers. With standard taper outer joint at the top for use with 10/18 or 10/30 jointed thermometers. Both lower inner joints are the same size. Side arm is at a 75¿ angle to the vertical tube. For thermometers see CG-3506-N.
Catalog No. CG107601 Supplier Chemglass Life Sciences Supplier No. CG107601
Adapter, Distilling, 24/40 - 10/30, 75mm Immersion, 75¿. Adapter is used to connect distilling column with vertical condensers. With standard taper outer joint at the top for use with 10/18 or 10/30 jointed thermometers. Both lower inner joints are the same size. Side arm is at a 75¿ angle to the vertical tube. For thermometers see CG-3506-N.