Learn More
Chemglass Life Sciences Distilling Adapter, Process Reactor, 45/50, Style "A", 14mm Stpk
SDP
Catalog No. 501311027
Encompass_Preferred
Distilling Adapter, Process Reactor, 45/50, Style "A", 14mm Stopcock. Similar to CG-1235, but with the lower stopcock replaced with a 0-14mm treaded valve having Perfluoro o-rings and a ¿¿ O.D. glass distillate takeoff for use with PFA compression tubing fittings. Adapter has two top outer joints and a lower inner. Adapter has a lower inner joint and two top outer joints.
Catalog No. 50-131-1027 Supplier Chemglass Life Sciences Supplier No. CG1235V21
Distilling Adapter, Process Reactor, 45/50, Style "A", 14mm Stopcock. Similar to CG-1235, but with the lower stopcock replaced with a 0-14mm treaded valve having Perfluoro o-rings and a ¿¿ O.D. glass distillate takeoff for use with PFA compression tubing fittings. Adapter has two top outer joints and a lower inner. Adapter has a lower inner joint and two top outer joints.