Learn More
Chemglass Life Sciences Distilling Adapter, Double, 10° Angled Side Neck, 29/42 Joint, 6mm PTFE Stopcock, 1/2" Swagelok
SDP
Catalog No. 501531064
Encompass_Preferred
Distilling Adapter, Double, 10 Angled Side Neck, 29/42 Joint, 6mm PTFE Stopcock, 1/2" Swagelok. Adapter is designed for use with flasks or reaction vessel lids that have a 10 angled side neck. Adapter has a lower inner joint and two top outer joints. Lower stopcock is a 6mm PTFE stopcock with a 1/2 or 3/4 OD distillate takeoff for use with a compression fitting and TFE tubing. PLEASE NOTE: ADAPTER CAN BE USED WITH COMPOUND ANGLED SIDE NECKS.
Catalog No. 50-153-1064 Supplier Chemglass Life Sciences Supplier No. CG123584
Distilling Adapter, Double, 10 Angled Side Neck, 29/42 Joint, 6mm PTFE Stopcock, 1/2" Swagelok. Adapter is designed for use with flasks or reaction vessel lids that have a 10 angled side neck. Adapter has a lower inner joint and two top outer joints. Lower stopcock is a 6mm PTFE stopcock with a 1/2 or 3/4 OD distillate takeoff for use with a compression fitting and TFE tubing. PLEASE NOTE: ADAPTER CAN BE USED WITH COMPOUND ANGLED SIDE NECKS.