Learn More
Chemglass Life Sciences Distilling Adapter, Double, 20° Angled Side Neck, 45/50 Joint, 10mm PTFE Stopcock, 3/4" Swagelok
SDP
Supplier:  Chemglass Life Sciences CG123589
Encompass_Preferred
Distilling Adapter, Double, 20 Angled Side Neck, 45/50 Joint, 10mm PTFE Stopcock, 3/4" Swagelok. Adapter is designed for use with flasks or reaction vessel lids that have a 20 angled side neck. Adapter has a lower inner joint and two top outer joints. Lower stopcock is a 10mm PTFE stopcock with a 1/2 or 3/4 OD distillate takeoff for use with a compression fitting and TFE tubing. PLEASE NOTE: ADAPTER CAN BE USED WITH COMPOUND ANGLED SIDE NECKS.
Catalog No. 50-153-1067