Learn More
Chemglass Life Sciences Distilling adapter, Double, 24/40 Joint, 6mm PTFE Stopcock, 1/2" Swagelock
SDP
Catalog No. CG123556
Encompass_Preferred
Distilling adapter, Double, 24/40 Joint, 6mm PTFE Stopcock, 1/2" Swagelock. Compact and simple distilling adapter for use with Chemglass process reactors. Vapor flows upward freely and distillate is collected in to the small bowl below the joint and through the stopcock into a distilling receiver. Adapter has a lower inner joint and is available with one or two top outer joints. Lower stopcock is either a 6mm or a 10mm PTFE with a 1/2¿ or 3/4¿ OD distillate takeoff for use with a compression fitting and TFE tubing. Adapter is 6¿ overall wide with joints offset 95mm center to center.
Catalog No. CG123556 Supplier Chemglass Life Sciences Supplier No. CG123556
Distilling adapter, Double, 24/40 Joint, 6mm PTFE Stopcock, 1/2" Swagelock. Compact and simple distilling adapter for use with Chemglass process reactors. Vapor flows upward freely and distillate is collected in to the small bowl below the joint and through the stopcock into a distilling receiver. Adapter has a lower inner joint and is available with one or two top outer joints. Lower stopcock is either a 6mm or a 10mm PTFE with a 1/2¿ or 3/4¿ OD distillate takeoff for use with a compression fitting and TFE tubing. Adapter is 6¿ overall wide with joints offset 95mm center to center.