Learn More
Chemglass Life Sciences Distilling Adapter, Process Reactor, 24/40, Style "B", 10°, 14mm Stpk
SDP
Catalog No. 5000980595
Encompass_Preferred
Similar to CG-1235, but with the lower stopcock replaced with a 0-14mm threaded valve having Perfluoro o-rings and a ½” O. D. glass distillate takeoff for use with PFA compression tubing fittings. Adapter has two top outer joints and a lower inner. Style “B” with lower angled joints are designed for use with flasks or reaction vessel lids that have a 10 degrees or 20 degrees angled side neck. Adapter has a lower inner joint and two top outer joints.
Catalog No. 50-009-80595 Supplier Chemglass Life Sciences Supplier No. CG1235V30
Similar to CG-1235, but with the lower stopcock replaced with a 0-14mm threaded valve having Perfluoro o-rings and a ½” O. D. glass distillate takeoff for use with PFA compression tubing fittings. Adapter has two top outer joints and a lower inner. Style “B” with lower angled joints are designed for use with flasks or reaction vessel lids that have a 10 degrees or 20 degrees angled side neck. Adapter has a lower inner joint and two top outer joints.