Learn More
Chemglass Life Sciences Distilling Bulb, 15mL, Kugelrohr, 14/20 Joint
SDP
Catalog No. NC1420902
Encompass_Preferred
Kugelrohr bulbs having 14/20 standard taper joints on both ends to connect to other bulbs or single neck round bottom flasks. CG-1012-14 adapter permits connection of Kugelrohr assembly to vacuum. For single neck 14/20 flasks see CG-1506.
Catalog No. NC1420902 Supplier Chemglass Life Sciences Supplier No. CG122711
Kugelrohr bulbs having 14/20 standard taper joints on both ends to connect to other bulbs or single neck round bottom flasks. CG-1012-14 adapter permits connection of Kugelrohr assembly to vacuum. For single neck 14/20 flasks see CG-1506.