Learn More
Chemglass Life Sciences Distilling Column, Hemple, Minum-Ware
SDP
Catalog No. 501532681
Encompass_Preferred
Distilling Column, Hemple, Minum-Ware. Hemple style column with indents just above the lower inner joint to support packing material. Top threaded outer joint and lower inner drip tip joint are 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).
Catalog No. 50-153-2681 Supplier Chemglass Life Sciences Supplier No. MW4501
Distilling Column, Hemple, Minum-Ware. Hemple style column with indents just above the lower inner joint to support packing material. Top threaded outer joint and lower inner drip tip joint are 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton O-ring (MW-100-08).