Learn More
Chemglass Life Sciences Distilling Column, Vigreux, Minum-Ware, Column Height 100, OAL 145
SDP
Catalog No. 50873059
Encompass_Preferred
Distilling Column, Vigreux, Minum-Ware, Column Height 100, OAL 145. Vigreux style column with indentations for improved vapor-liquid contact. Column height is the length of the indented section. Upper threaded outer joint and lower inner joint are 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08).
Catalog No. 50-873-059 Supplier Chemglass Life Sciences Supplier No. MW5202
Distilling Column, Vigreux, Minum-Ware, Column Height 100, OAL 145. Vigreux style column with indentations for improved vapor-liquid contact. Column height is the length of the indented section. Upper threaded outer joint and lower inner joint are 14/10. Supplied complete with open-top screw cap (MW-102-33) and Viton o-ring (MW-100-08).